Skip to Content
Merck
Phorbol 12,13-dibutyrate ≥98% (TLC), powder

Phorbol 12,13-dibutyrate

Phorbol 12,13-dibutyrate ≥98% (TLC), powder
Synonym(s):
PDBu, Phorbol-12,13-dibutyrate
Empirical Formula (Hill Notation):
C28H40O8
CAS Number:
Molecular Weight:
504.61

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing