Skip to Content
Merck
All Photos(1)

Key Documents

CRM05691

Supelco

Anilazine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

6-(2-Chloroanilino)-2,4-dichloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5Cl3N4
CAS Number:
Molecular Weight:
275.52
Beilstein:
223133
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

Clc1nc(Cl)nc(Nc2ccccc2Cl)n1

InChI

1S/C9H5Cl3N4/c10-5-3-1-2-4-6(5)13-9-15-7(11)14-8(12)16-9/h1-4H,(H,13,14,15,16)

InChI key

IMHBYKMAHXWHRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. The CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Anilazine is a triazine compound, used as a non-systemic foliar fungicide on crops such as vegetables, cereals, ornamental plants, fruits, coffee, and tobacco.
As per the commission regulation no 1107/2009, repealing the council directive 91/414/EEC, anilazine is not approved for use in European Union. A default maximum residue limit (MRL) of 0.01 mg/kg is applied for the presence of anilazine in frozen/fresh fruits and tree nuts as per regulation (EC) no. 899/2012.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The CRM may also find the following uses:

  • Analysis of long-term persistence of three 14C-labeled pesticides- ethidimuron, methabenzthiazuron, anilazine, and their metabolites in soil
  • Gas chromatographic-mass spectrometric (GC-MS) determination of different classes of pesticide residues from various wastewater samples obtained from culturing and processing of potatoes

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolai D Jablonowski et al.
Environmental pollution (Barking, Essex : 1987), 168, 29-36 (2012-05-18)
The fate of the 14C-labeled herbicides ethidimuron (ETD), methabenzthiazuron (MBT), and the fungicide anilazine (ANI) in soils was evaluated after long-term aging (9-17 years) in field based lysimeters subject to crop rotation. Analysis of residual 14C activity in the soils
DETERMINATION OF THE PESTICIDES IN WATER USED IN THE CULTURE AND PROCESSING OF POTATOES
Ribeiro P H, et al.
Quimica nova, 1026- 1034 (2020)
Nicolai D Jablonowski et al.
Environmental pollution (Barking, Essex : 1987), 168, 29-36 (2012-05-18)
The fate of the 14C-labeled herbicides ethidimuron (ETD), methabenzthiazuron (MBT), and the fungicide anilazine (ANI) in soils was evaluated after long-term aging (9-17 years) in field based lysimeters subject to crop rotation. Analysis of residual 14C activity in the soils

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service