Z768626
Nalgene® square graduated media bottle
without cap, PET, sterile, capacity 125 mL
Synonym(s):
media storage bottle, square plastic bottle
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
PET
square bottle
Joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations
without cap
packaging
pack of 96 ea (4 x pack of 24)
manufacturer/tradename
Nalgene 842044-0125
capacity
125 mL
General description
Nalgene® Square PET media bottles offer sterility, lightweight construction, and shatter resistance. They are specifically designed for use in filling lines.
Features and Benefits
- Has molded-in graduations and neck-ring.
- Non-cytotoxic, non-hemolytic, and non-pyrogenic.
- Exhibits exceptional gas barrier properties.
- Gamma irradiated to 10^-6 to achieve sterility assurance level (SAL).
- Composed of material free from ADC (animal derived component).
Other Notes
Closures for these bottles are sold separately.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)