Skip to Content
Merck
All Photos(1)

Key Documents

EP0030108094

Eppendorf® Protein LoBind tubes

PCR clean, withstand, 18,000 x g, capacity 0.5 mL, pack of 100 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

cap (plug seal)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube

sterility

non-sterile

feature

PCR clean
graduations
neck thread Push fit

packaging

pack of 100 ea

manufacturer/tradename

Eppendorf® 0030108094

technique(s)

PCR: suitable

capacity

0.5 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Eppendorf® LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • Available in tube formats for easy-up scaling.
  • Precise lid sealing to minimize evaporation.

Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service