Skip to Content
Merck
All Photos(1)

Key Documents

EP0030010043

Eppendorf® epT.I.P.S.® G Singles, Biopur®

volume (2-200 μL), L (53 mm), yellow Tray, pkg of 100 tips, individually wrapped

Synonym(s):

Eppendorf G Singles, Eppendorf Pipette Tips, epT.I.P.S. G Singles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
L:
(53 mm)
maximum volume:
200 μL
volume:
(2-200 μL)
sterility:
non-sterile
feature:
individually wrapped
packaging:
pack of 100 ea (individually wrapped)
pkg of 100 tips
manufacturer/tradename:
Eppendorf® 0030010043

material

colorless

sterility

non-sterile

feature

individually wrapped

packaging

pack of 100 ea (individually wrapped)
pkg of 100 tips

manufacturer/tradename

Eppendorf® 0030010043

L

(53 mm)

maximum volume

200 μL

volume

(2-200 μL)

color

yellow Tray

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® totally integrated pipetting system (epT.I.P.S.) pipette tips fit perfectly with the Eppendorf pipette to form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. epT.I.P.S pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all the specific needs of your samples. This results in reduced-force tip attachment and ejection with complete sealing. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials.

Application

Eppendorf® epT.I.P.S.® G Singles, Biopur® is suitable for:
  • pipetting liquids
  • Dispensing liquids
  • Mixing liquids
  • Filling plates and reaction vessels
  • Filling gels for electrophoresis
  • Phase extraction and removal of supernatants

Features and Benefits

  • Eppendorf® Biopur® pipette tips are PCR clean, sterile, pyrogen-, ATP- and DNA-free
  • For use in the pharmaceutical and food industry, molecular biology, forensic and cell technology
  • Batch number and expiration date on each blister package
  • The nose cone design allows epT.I.P.S. pipette tips to also be used universally, including pipette models from other manufacturers
  • The ergonomically optimized cone geometry provides the optimal pipette fit on the Eppendorf pipette
  • Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
  • Fine retracted or extended tips for pipetting small volumes easily

Other Notes

Replaces order no. EP0030010035.

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service