Skip to Content
Merck
All Photos(1)

Documents

158801

Sigma-Aldrich

Tetrachlorophthalonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Cl4-1,2-(CN)2
CAS Number:
Molecular Weight:
265.91
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

98%

form

powder, crystals or chunks

mp

249-252 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Clc1c(Cl)c(Cl)c(C#N)c(C#N)c1Cl

InChI

1S/C8Cl4N2/c9-5-3(1-13)4(2-14)6(10)8(12)7(5)11

InChI key

OQHXZZGZASQSOB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Photochemistry with donor-acceptor systems: part 2. Photoreaction of tetrachlorophthalonitrile with naphthalene
A.C. Pratt and F.M. Grimley
Proc. Royal Soc. Lond. B. (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service