Skip to Content
Merck
All Photos(1)

Key Documents

GFCP000800

Millipore

Glass Fiber Conjugate Pad 8mm x 100m Roll

Lateral Flow, Assay Development, lateral flow immunoassays, lateral flow assay device, Immunoassay IVDs

Synonym(s):

Glass fiber pad 8 mm width roll

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.23

Quality Level

packaging

pkg of 1 roll

manufacturer/tradename

SureWick®

roll L

100 m (328 ft)

roll W

8 mm , slit width

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service