Skip to Content
Merck
All Photos(1)

Key Documents

900131E

Avanti

13(S)HODE Ethanolamide

Avanti Research - A Croda Brand

Synonym(s):

13S-Hydroxy-N-(2-hydroxyethyl)-9Z,11E-octadecadienamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37NO3
CAS Number:
Molecular Weight:
339.51
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

ethanol solution

packaging

pkg of 1 × 1 mg (900131E-1mg)
pkg of 1 × 500 μg (900131E-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (900131E-1MG)
500 μg/mL (900131E-500UG)

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−70°C

Packaging

2ml Amber Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service