Skip to Content
Merck
All Photos(2)

Key Documents

830841P

Avanti

06:0 PA

1,2-dihexanoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1,2-dicaproyl-sn-glycero-3-phosphate (sodium salt); PA(6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O8PNa
CAS Number:
Molecular Weight:
390.34
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (830841P-10mg)
pkg of 1 × 25 mg (830841P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830841P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCC)=O)COC(CCCCC)=O.[Na+]

InChI

1S/C15H29O8P.Na/c1-3-5-7-9-14(16)21-11-13(12-22-24(18,19)20)23-15(17)10-8-6-4-2;/h13H,3-12H2,1-2H3,(H2,18,19,20);/q;+1/p-1/t13-;/m1./s1

InChI key

RSKVQVAHZSAALH-BTQNPOSSSA-M

Application

06:0 PA has been used for flotation assay.

Packaging

5 mL Clear Glass Sealed Ampule (830841P-10mg)
5 mL Clear Glass Sealed Ampule (830841P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assay to Measure Interactions between Purified Drp1 and Synthetic Liposomes
Adachi Y, et al.
Bio-protocol, 7(9) (2017)
Yasunori Watanabe et al.
Nature communications, 6, 7922-7922 (2015-08-04)
Eukaryotic cells are compartmentalized into membrane-bounded organelles whose functions rely on lipid trafficking to achieve membrane-specific compositions of lipids. Here we focused on the Ups1-Mdm35 system, which mediates phosphatidic acid (PA) transfer between the outer and inner mitochondrial membranes, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service