Merck
All Photos(4)
Key Documents
Z513334
Micro spatulas
flat rounded/tapered PTFE coated ends, L 7 5/8 in. (190 mm)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Pricing and availability is not currently available.
material
nickel-stainless steel
packaging
pkg of 1 ea
L
7 5/8 in. (190 mm)
General description
Nickel stainless, PTFE coated spatula with one rounded end and one tapered end. Rounded end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long. Tapered end approximately 1/4 in. (6 mm) wide x 1 7/8 in. (48 mm) long.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?