Key Documents
SLW1519/12
Pyrex® Media-Lab Bottles, amber glass, with-out cap and pouring ring, with printed trace code
Synonym(s):
Pyrex, SciLabware 1519/12
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
round amber glass bottle
Joint
Joints threaded neck (GL45)
sterility
non-sterile
feature
graduations
without cap
with printed trace code
without pouring ring
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1519/12
H
260 m
body diam.
136 mm
General description
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Manufactured from amber-coated borosilicate glass
  • UV absorbent up to 500 nm
  • Ideal for storage of light sensitive materials
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)