Skip to Content
Merck
All Photos(1)

Key Documents

GF97831996

Nitinol alloy, Ni55Ti45

1000mm tube, 1.27mm outside diameter, 0.15mm wall thickness, 0.97mm inside diameter, superelastic

Synonym(s):

NI207130

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

Pricing and availability is not currently available.

form

tubes

manufacturer/tradename

Goodfellow 978-319-96

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
555299346550194
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

neat

format

neat

format

neat

format

neat

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

assay

≥98.0% (HPLC)

assay

≥98.0% (GC)

assay

≥98.0% (HPLC)

assay

≥97.5% (HPLC)

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service