Key Documents
BR704534
BRAND® filling tube
for seripettor®, L 500 mm
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
UNSPSC Code:
41121800
NACRES:
NB.01
material
polypropylene
description
autoclavable for sterile applications
packaging
pack of 2 ea
manufacturer/tradename
BRAND 704534
L
500 mm
Related Categories
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)