BAB617003300
SP Bel-Art® Durac® 4-Channel Electronic Timer and Clock
with certificate of calibration
All Photos(1)
About This Item
UNSPSC Code:
41112303
NACRES:
NB.01
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B61700-3300
measuring range
accuracy: ±3 sec, resolution: 0.01
H × W
80 mm (3.1 in.) × 80 mm (3.1 in.)
General description
This 4-channel timer has a capacity of 99 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
Features and Benefits
- Counts either up or down; remembers the last time set
- Lay flat on a bench, use the flip-open stand for a vertical view, or carry it on a lab coat with the convenient pocket clip
- Requires one AAA battery (included)
- Features a Certificate of Calibration and individual serial number, these electronic timers are tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)