BAB605060000
SP Bel-Art® H-B Enviro-Safe® General Purpose Liquid-In-Glass Thermometer
temp -35-50 °C, immersion level, Total Immersion
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
graduation 1 °C
immersion level Total Immersion
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60506-0000
temp measuring range
-35-50 °C
accuracy: ±0.1 °C
L
12 in. (300 mm)
Looking for similar products? Visit Product Comparison Guide
General description
Earth friendly, EnviroKleen certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass.
Features and Benefits
Earth friendly, EnviroKleen certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass.
- Meet government, education, and industry mandates to purchase “Green” products
- Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries
- Manufactured and tested by our USA quality team
- Individually serialized
- Traceable to NIST
- Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Enviro-Safe is a registered trademark of H-B Instrument Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service