933481
Wafer tweezer
Style 45WF, PTFE Coated
Synonym(s):
Wafer Tweezers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
EMS techware
Specificity
Handling 3", top fingers, stepped bottom paddle. PTFE coated.
Length: 130mm
Length: 130mm
Application
Wafer tweezer, Style 45WF, PTFE Coated are used for handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips. Made from antimagnetic/anti-acid steel with smooth surface.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service