615943
Fmoc-Leu-OH-5,5,5-d3
99 atom % D
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-leucine-5,5,5-d3, L-Leucine-5,5,5-d3, N-Fmoc
About This Item
Recommended Products
isotopic purity
99 atom % D
Assay
99% (CP)
optical activity
[α]20/D -25°, c = 0.5 in DMF
technique(s)
bio NMR: suitable
mp
152-156 °C (lit.)
application(s)
peptide synthesis
functional group
Fmoc
mass shift
M+3
SMILES string
[2H]C([2H])([2H])C(C)C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O
InChI
1S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1/i1D3/t13?,19-
InChI key
CBPJQFCAFFNICX-VWCYLIGHSA-N
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service