L-15 Media Formulation
Component | L1518 [1x] g/L | L4386 [1x] g/L | L5520 g/L |
---|---|---|---|
Inorganic Salts | |||
Calcium Chloride | 0.1396 | 0.1396 | 0.1396 |
Magnesium Chloride | 0.0937 | 0.0937 | 0.0937 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 0.4 | 0.4 |
Potassium Phosphate Monobasic (anhydrous) | 0.06 | 0.06 | 0.06 |
Sodium Chloride | 8 | 8 | 8 |
Sodium Phosphate Dibasic (anhydrous) | 0.19 | 0.19 | 0.19 |
Amino Acids | |||
L-Alanine | 0.225 | 0.225 | 0.225 |
L-Arginine (free base) | 0.5 | 0.5 | 0.5 |
L-Asparagine (anhydrous) | 0.25 | 0.25 | 0.25 |
L-Cysteine (free base) | 0.12 | 0.12 | 0.12 |
L-Glutamine | 0.3 | 0.3 | — |
Glycine | 0.2 | 0.2 | 0.2 |
L-Histidine | 0.25 | 0.25 | 0.25 |
L-Isoleucine | 0.125 | 0.125 | 0.125 |
L-Leucine | 0.125 | 0.125 | 0.125 |
L-Lysine | 0.0937 | 0.0937 | 0.0937 |
L-Methionine | 0.075 | 0.075 | 0.075 |
L-Phenylalanine | 0.125 | 0.125 | 0.125 |
L-Serine | 0.2 | 0.2 | 0.2 |
L-Threonine | 0.3 | 0.3 | 0.3 |
L-Tryptophan | 0.02 | 0.02 | 0.02 |
L-Tyrosine (free base) | 0.3 | 0.3 | 0.3 |
L-Valine | 0.1 | 0.1 | 0.1 |
Vitamins | |||
Choline Chloride | 0.001 | 0.001 | 0.001 |
Flavin Mononucleotide • Na | 0.0001 | 0.0001 | 0.0001 |
Folic Acid | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.001 | 0.001 |
DL-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 | 0.001 |
Pyridoxine • HCl | 0.001 | 0.001 | 0.001 |
Thiamine Monophosphate • HCl | 0.001 | 0.001 | 0.001 |
Other | |||
D-Galactose | 0.9 | 0.9 | 0.9 |
Phenol Red • Na | 0.011 | 0.011 | 0.011 |
Pyruvic Acid • Na | 0.55 | 0.55 | 0.55 |
Add | |||
L-Glutamine | — | — | 0.3 |
예기치 못한 오류가 발생했습니다.
Network error: Failed to fetch