Merck
HomeMammalian Cell CultureAntifungals Reagents in Cell Culture
Antifungals Reagents in Cell Culture
Fungal Contamination in Cell Culture
Fungal infections in cell cultures can have severe consequences for cell health and experimental reproducibility. Fortunately, these infections are often visible to the naked eye. Fungal contaminations change the color of phenol red and can cause rapid on-set of turbidity in the cell culture media. While testing cultures for fungi is an important part of routine quality control, it is important to eliminate and prevent fungal infections before they start.
Antifungals for Cell Culture
Antifungal compounds are chemicals that target the integrity of fungal cell wall or cell membrane, thereby controlling or eliminating fungal infections in cell cultures. Most antifungals either directly or indirectly increase the porosity of the fungi cell wall or membrane. These compounds can also interfere with the steps in cell wall synthesis. In addition to antibacterials, antifungals have been evolved as critical additives to culture medium when culturing cell lines.
Amphotericin B
Amphotericin B is also known as Fungizone and is most effective against fungi and yeast infections. The compound binds to sterols and induces formation of pores in the membrane. These pores allow small molecules to leak out, thereby destroying the infection. Amphotericins are stable in cell culture for 3 days at 37˚C.
Product #
Name
Form
Recommended Concentration
E3789
Amphotericin B low endotoxin
Powder
2.5 mg/mL
A2411
Amphotericin B from Streptomyces sp.
Solid
2.5 mg/mL
A9528
Amphotericin B solubilized
Solubilized powder
5.6 mg/mL
A2942
Amphotericin B solution
Liquid
10 ml/L
Nystatin
The antifungal Nystatin is also known as Mycostatin or Fungicidine. Like Amphotericin B, Nystatin eliminates fungal infections by binding to the sterols in the fungal cell membrane and causing small molecules to leak out. However, compared to Amphotericin B, Nystatin has a larger concentration window and is less toxic to the cells in culture. Nystatins are stable in cell culture for 3 days at 37 ˚C.
We also offer water-soluble versions of Nystatin (N6803 & N9150) which are convenient to use, avoid the use of organic solvents, and are most suitable to protect sensitive cells against contamination. 
Product #
Name
Form
Recommended Concentration
N6803
Nystatin water soluble
Solubilized powder
3-5 mg/mL
N9150
Nystatin Ready made solution
Liquid
3-5 mg/mL
N6261
Nystatin
Powder
2.5 mg/mL
N4014
Nystatin
Powder
50 mg/mL
N1638
Nystatin Suspension
Suspension
24 ml/L
Other Antifungals for Cell Culture
Product #
Name
Form
A5955
Antibiotic Antimycotic Solution (100×), Stabilized
Suspension
T5535
Thiabendazole for plant cell culture
Powder
P8556
Pentachloronitrobenzene for plant cell culture
Powder
H3274
Hygromycin B (for insect cell culture)
Powder
H9773
Hygromycin B (for plant cell culture)
Powder
B8416Bleomycin sulfate from Streptomyces verticillusSolid
Other Sources of Contamination
Contamination is a significant concern in cell culture experiments and gathering reliable data. Mycoplasma contamination, bacterial contamination, and other common contaminants can negatively affect data reproducibility. Therefore, it is important to protect your cultures by exploring antibiotic powders and solutions for your cells, selecting the right antibiotics for your cells, and determining a suitable antibiotic concentration for your cells. 
Products
예기치 못한 오류가 발생했습니다.
Network error: Failed to fetch