HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18
Figure 1.
Conditions | |
---|---|
Column | SUPELCOSIL LC-18, 25 cm x 4.6 mm I.D., 5 µm particles (58298) |
Mobile Phase | [A] acetonitrile; [B] acetone; (59.0:41.0, v/v, A:B) |
Flow Rate | 1 mL/min |
Injection | 10 µL of 9% C8 to C18 saturated and unsaturated FAMEs in mobile phase |
Detector | RI |
Description | |
---|---|
Categories | Analytical Chromatography, Fatty Acids, FAMEs |
Featured Industry | Chemicals and Industrial Polymers Food and Beverages |
Legal Information | SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC |
Suitability | Application for HPLC |
Materials
예기치 못한 오류가 발생했습니다.
Network error: Failed to fetch