Merck
HPLC Analysis of Lorazepam Enantiomers on Astecยฎ CHIROBIOTICยฎ T
HPLC Analysis of Lorazepam Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
๋ถ„์„ ์ปฌ๋Ÿผ
์ œํ’ˆ ๋ฒˆํ˜ธ
์„ค๋ช…
๊ฐ€๊ฒฉ
Astecยฎ CHIROBIOTICยฎ T Chiral HPLC Column
5 μm particle size, L ร— I.D. 25 cm ร— 4.6 mm
ํ‘œ์ค€ ๋ฌผ์งˆ
์ œํ’ˆ ๋ฒˆํ˜ธ
์„ค๋ช…
๊ฐ€๊ฒฉ
(ยฑ)-Lorazepam
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
methanol
flow rate
1 mL/min
column temp.
25 ยฐC
detector
UV, 230 nm
injection
1 μL
sample
5 mg/mL in methanol
์„ค๋ช…
๋ฒ•์  ์ •๋ณด