Merck
HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T (No Additives)
HPLC Analysis of Methionine Enantiomers on Astec® CHIROBIOTIC® T (No Additives) application for HPLC
Materials
분석 컬럼
제품 번호
설명
가격
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
표준 물질
제품 번호
설명
가격
DL-Methionine
≥99%
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] acetonitrile; (30:70, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm