Merck
HPLC Analysis of Dehydroascorbic Acid and Ascorbic Acid (Vitamin C) on SeQuant® ZIC-HILIC
HPLC Analysis of Dehydroascorbic Acid and Ascorbic Acid (Vitamin C) on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate
0.5 mL/min
pressure
375 psi (2.6 MPa)
detector
240 nm
injection
5 μL
설명
법적 정보
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany