콘텐츠로 건너뛰기
Merck
  • Structure-activity studies of antitumor agents based on pyrrolo[1,2-a]benzimidazoles: new reductive alkylating DNA cleaving agents.
Structure-activity studies of antitumor agents based on pyrrolo[1,2-a]benzimidazoles: new reductive alkylating DNA cleaving agents.
Journal of medicinal chemistry (1991-10-01)
I Islam, E B Skibo, R T Dorr, D S Alberts
초록
Described herein are structure-activity studies of new antitumor agents based on the pyrrolo[1,2-a]benzimidazole (PBI) ring system. These compounds were designed as new DNA cross-linkers mimicking the mitomycin antitumor agents. Actually, the PBI derivatives were found to have anthracycline-like features: (i) shared cross resistance with doxorubicin in a human myeloma line, (ii) cardiotoxicity, and (iii) excellent DNA strand cleaving capability. The DNA strand cleavage is thought to result from reductive alkylation of DNA followed by the generation of reactive oxygen radicals. The best antitumor agent studied is 6-N-aziridinyl-3-hydroxy-7-methyl-2,3-dihydro-1H-pyrrolo- [1,2-a]benzimidazole-5,8-dione 3-acetate (PBI-A), which possesses nanomolar IC50 values against various human ovarian and colon cancer cell lines.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Potassium nitrosodisulfonate