콘텐츠로 건너뛰기
Merck
  • Synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
Synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.
Journal of medicinal chemistry (1986-08-01)
A N Brubaker, M Colley
초록
Three 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes that contained either the benzyl, 3-indolylmethyl, or 4-indolylmethyl group at the 6-position were synthesized via alkylation of the pyrrolidine enamine of the key intermediate, ethyl 3-oxopiperidine-1-carboxylate. The spirodecane derivatives were evaluated for in vivo central and peripheral dopamine agonist activity. None of the compounds displayed central nervous system activity; however, the 4-indolymethyl analogue exhibited potent dopamine agonist activity in the cat cardioaccelerator nerve assay and possesses an ID50 of 0.095 mumol/kg compared to apomorphine, which possesses an ID50 of 0.0348 mumol/kg in the same assay.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Methyl 2-methyl-3-nitrobenzoate, 97%
가격 및 재고 정보를 현재 이용할 수 없음