콘텐츠로 건너뛰기
Merck
  • Large-area graphene films by simple solution casting of edge-selectively functionalized graphite.
Large-area graphene films by simple solution casting of edge-selectively functionalized graphite.
ACS nano (2011-05-20)
Seo-Yoon Bae, In-Yup Jeon, Jieun Yang, Noejung Park, Hyeon Suk Shin, Sungjin Park, Rodney S Ruoff, Liming Dai, Jong-Beom Baek
초록
We report edge-selective functionalization of graphite (EFG) for the production of large-area uniform graphene films by simply solution-casting EFG dispersions in dichloromethane on silicon oxide substrates, followed by annealing. The resultant graphene films show ambipolar transport properties with sheet resistances of 0.52-3.11 kΩ/sq at 63-90% optical transmittance. EFG allows solution processing methods for the scalable production of electrically conductive, optically transparent, and mechanically robust flexible graphene films for use in practice.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
4-Ethylbenzoic acid, 99%