콘텐츠로 건너뛰기
Merck
  • Dietary administration of zingerone to enhance growth, non-specific immune response, and resistance to Vibrio alginolyticus in Pacific white shrimp (Litopenaeus vannamei) juveniles.
Dietary administration of zingerone to enhance growth, non-specific immune response, and resistance to Vibrio alginolyticus in Pacific white shrimp (Litopenaeus vannamei) juveniles.
Fish & shellfish immunology (2011-12-17)
Yueh-Ping Chang, Chun-Hung Liu, Chih-Chung Wu, Chi-Ming Chiang, Juang-Lin Lian, Shu-Ling Hsieh
초록
Zingerone, one of the active components of ginger, is a phenolic alkanone with antioxidant and anti-inflammatory properties. The effects of zingerone supplementation on the growth, immunity, and disease resistance of Pacific white shrimp (Litopenaeus vannamei) juveniles were studied. Four experimental diets, including a control diet (without zingerone enrichment) and 1, 2.5, and 5 mg zingerone (kg diet)(-1) were used. After 56 days of culture, shrimp fed diets supplemented with 1, 2.5, and 5 mg zingerone (kg diet)(-1) had significantly greater weight gain and feed efficiency than the controls. Furthermore, after 56 days of culture, shrimp fed all doses of the zingerone diet had higher survival rates compared to the controls after 24-72 h of challenge by the pathogen, Vibrio alginolyticus. Significantly increased phenoloxidase levels were found in shrimp fed the zingerone diets at all doses, and respiratory bursts, lysozyme and phagocytic activities of shrimp fed 2.5 and 5 mg zingerone (kg diet)(-1) also significantly increased. Neither the total hemocyte count nor superoxide dismutase activity of the experimental and control groups revealed significant differences at any dose. The results indicate that zingerone can be recommended as a supplement to shrimp feed to increase growth, immunity, and disease resistance against the pathogen, V. alginolyticus. Use of zingerone as appetizer and immunostimulant in shrimp is promising.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Vanillylacetone, ≥96%, FG
가격 및 재고 정보를 현재 이용할 수 없음