콘텐츠로 건너뛰기
Merck
  • Erythema multiforme after contact dermatitis in response to an epoxy sealant.
Erythema multiforme after contact dermatitis in response to an epoxy sealant.
Journal of the American Academy of Dermatology (1991-08-01)
M J Whitfeld, J K Rivers
초록
A case of erythema multiforme associated with an allergic contact dermatitis in response to an epoxy-based compound is presented. Patch tests revealed a positive reaction to both the epoxy resin and the hardener. Chemicals applied directly to the skin should be considered as a potential cause of erythema multiforme.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
5-Amino-1,3,3-trimethylcyclohexanemethylamine, mixture of cis and trans, ≥99%