콘텐츠로 건너뛰기
Merck
  • A New IL6 Isoform in Chinese Soft-Shelled Turtle (Pelodiscus sinesis) Discovered: Its Regulation during Cold Stress and Infection.
A New IL6 Isoform in Chinese Soft-Shelled Turtle (Pelodiscus sinesis) Discovered: Its Regulation during Cold Stress and Infection.
Biology (2020-05-30)
Zuobing Zhang, Miao Tian, Ruxin Song, Xiao Xing, Yong Fan, Lan Wang, Cuijuan Niu, Roy A Dalmo
초록
The Chinese soft-shelled turtle (Pelodiscus sinesis) is a widely cultured commercial species in East and Southeast Asian countries. The turtles frequently suffer from acute cold stress during farming in China. Stress-induced factor such as Interleukin-6 (IL6) is a multifunctional molecule that plays important roles in innate and adaptive immune response. In the present study, we found that the turtle possessed two IL6 transcripts, where one IL6 transcript contained a signal peptide sequence (psIL6), while the other IL6 transcript (psIL6ns) possessed no such signal peptide gene. To test any differential expression of the two isoforms during temperature and microbial stress, turtles were adapted to optimal environmental water temperature (25 °C), stressed by acute cooling for 24 h, followed with the challenge of Aeromonas hydrophila (1.8 × 108 CFU) or Staphylococcus aureus (5.8 × 108 CFU). Gene characterization revealed that psIL6ns, a splicer without codons encoding a signal peptide and identical to the one predicted from genomic sequence, and psIL6, a splicer with codons encoding a signal peptide, were both present. Inducible expression was documented in primary spleen cells stimulated with ConA and poly I: C. The splenic and intestinal expression of psIL6ns and psIL6 was increased in response to temperature stress and bacterial infection.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder