콘텐츠로 건너뛰기
Merck
  • Nonredundant functions for Akt isoforms in astrocyte growth and gliomagenesis in an orthotopic transplantation model.
Nonredundant functions for Akt isoforms in astrocyte growth and gliomagenesis in an orthotopic transplantation model.
Cancer research (2011-04-22)
Raelene Endersby, Xiaoyan Zhu, Nissim Hay, David W Ellison, Suzanne J Baker
초록
The AKT family, comprising three highly homologous kinases, is an essential mediator of the PTEN/PI3K pathway, which is deregulated in many human cancers. A thorough understanding of the specific activities of each isoform in normal and disease tissues is lacking. We evaluated the role of each Akt isoform in gliomagenesis by using a model system driven by common glioma abnormalities, loss of function of p53 and Pten, and expression of EGFRvIII. Both Pten deletion and EGFRvIII expression accelerated the proliferation of p53-null primary murine astrocytes. All three Akt isoforms were expressed and phosphorylated in astrocytes, with significantly higher activation in Pten-null cells. Despite substantial compensation in many contexts when individual Akt isoforms were inhibited, isoform-specific effects were also identified. Specifically, loss of Akt1 or Akt2 decreased proliferation of Pten wild-type astrocytes, whereas combined loss of multiple isoforms was needed to inhibit proliferation of Pten-null astrocytes. In addition, Akt3 was required for anchorage-independent growth of transformed astrocytes and human glioma cells, and Akt3 loss inhibited invasion of transformed astrocytes. EGFRvIII expression transformed p53-null astrocytes with or without Pten deletion, causing rapid development of high-grade astrocytoma on intracranial transplantation. Furthermore, tumorigenesis of Pten;p53-null astrocytes expressing EGFRvIII was delayed by Akt1 loss and accelerated by Akt2 loss. Taken together, these results indicate context-dependent roles for individual Akt isoforms and suggest that there may be heterogeneous tumor response to isoform-specific inhibitors.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Akt3/PKBγ Antibody, Upstate®, from rabbit