주요 문서
G1006856
UHPLC/MS Analysis of Insulin Variants and Analogs on BIOshell A160 Peptide C18 Using Difluoroacetic Acid as a Mobile Phase Modifier
application for UHPLC
로그인조직 및 계약 가격 보기
About This Item
기술
UHPLC: suitable
검사 파라미터
column: BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase: [A] 75:25 water (0.1% v/v difluoroacetic acid): acetonitrile (0.1% v/v difluoroacetic acid); [B] 50:50 water (0.1% v/v difluoroacetic acid): acetonitrile (0.1% v/v difluoroacetic acid); (77:23, A:B)
flow rate: 0.2 mL/min
column temp.: 75 °C
injection: MS, ESI(+) TOF, TIC 100-3000 m/z
injection: 0.5 μL
sample: Mixture of six insulin variants, 100 μg/mL, water (0.1% v/v difluoroacetic acid)
적합성
application for UHPLC
응용 분야
life science and biopharma
분석 메모
BIOshell A160 Peptide C18 is an ideal column for reversed-phase separations of small proteins to large peptide sequences. There is over 75% sequence similarity between all six of the insulin variants in the sample; therefore, separation of these variants is achieved through the subtle differences in the polarities of surface residues on the peptides. The use of difluoroacetic acid (DFA) has less of an effect on ion suppression than trifluoroacetic acid (TFA), thus allowing for mass spectral detection of the insulin variants.
법적 정보
분석물
설명
    insulin-Glargine
    insulin-Bovine
    insulin-Asp
    insulin-Lispro
    insulin-Human
    insulin-Porcine
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)