기술
HPLC: suitable
검사 파라미터
column: Zenix SEC-300 (2 ea), 30 cm x 7.8 mm I.D., 3 μm partcles, 300 Å, connected in tandem (Z777033)
mobile phase: 150 mM Sodium Phosphate Buffer pH=7.0
flow rate: 0.5 mL/min
column temp.: 25 °C
detector: Refractive Index at 30 °C
injection: 10 μL
sample: 1.PEG 81,400 Da (2mg/mL)
2.PEG 50,630 Da (2mg/mL)
3.PEG 40,000 Da (2mg/mL)
4.PEG 26,100 Da (2mg/mL)
5.PEG 8,730 Da (2mg/mL)
적합성
application for HPLC
응용 분야
life science and biopharma
일반 설명
Pegylation of biotherapeutic drugs improves solubility, increases half life in the blood stream and reduces immunogenicity. Size exclusion chromatography (SEC) is often applied to characterize protein–polyethylene glycol (PEG) conjugates and establish the number of attached PEG chains, since SEC is a precise and robust technique that combines ease of set up and strainghtforward method develpopment.
This chromatogram shows the separation of four polyethyleneglycol polymers varying in molecular mass 8730 to 81,400 Da on two Zenix® SEC-300 columns in series.
This chromatogram shows the separation of four polyethyleneglycol polymers varying in molecular mass 8730 to 81,400 Da on two Zenix® SEC-300 columns in series.
법적 정보
Zenix is a registered trademark of Sepax Technologies
가장 최신 버전 중 하나를 선택하세요:
자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..
고객지원팀으로 연락바랍니다.