주요 문서
56651-U
Supelco
HyperShear Ultra HPLC High Pressure Static Mixer
Binary tee housing assembly, volume 250 μL
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115854
material
(wetted material 316 stainless steel/PEEK)
설명
Port size, 10-32 CPI (standard HPLC fittngs for 1/16" tubing)
Pressure rated to 18,000 psi
용적
250 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Raffeal Bennett et al.
Analytica chimica acta, 999, 161-168 (2017-12-20)
The value of exploring selectivity and solvent strength ternary gradients in enhanced fluidity liquid chromatography (EFLC) is demonstrated for the separation of inulin-type fructans from chicory. Commercial binary pump systems for supercritical fluid chromatography only allow for the implementation of