문서
502618
Upchurch Microtight® Tubing Sleeves
I.D. 0.013 in. (330 μm), orange
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115700
material
orange
포장
pkg of 10 ea
I.D.
0.013 in. (330 μm)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Use these PEEK sleeves with all Microtight fittings, to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.
추천 제품
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
법적 정보
Microtight is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
시험 성적서(COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact 고객 지원 부서