주요 문서
28076-U
Supelco
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 3.00 μm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12000000
eCl@ss:
32119290
NACRES:
SB.54
material
fused silica
Agency
ASTM® D1945,D3710,D4815,D5442,D5580,D7096
EN 14110
EPA 8015,TO-17
GPA 2186,2286
NIOSH 1403,1500/1501,2542
OSHA 53,56
meets requirements for USP G1, G2, and G9
파라미터
-60-260 °C temperature (isothermal)
-60-280 °C temperature (programmed)
베타 값
44
df
3.00 μm
기술
gas chromatography (GC): suitable
길이 × I.D.
30 m × 0.53 mm
기질 활성군
Bonded; poly(dimethyl siloxane) phase
응용 분야
agriculture
biofuels
chemicals and industrial polymers
cleaning products
clinical
cosmetics
flavors and fragrances
food and beverages
forensics and toxicology
industrial hygiene
personal care
petroleum
컬럼 유형
capillary non-polar
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Application: This column is designed for general purpose applications where a non-polar column is required. Analytes will be separated primarily according to boiling point.
USP Code: This column meets USP G1, G2, and G9 requirements.
Phase:
  • Bonded
  • Poly(dimethyl siloxane)
Temp. Limits:
  • ≤0.32 mm I.D., <2 μm: -60 °C to 325 °C (isothermal) or 350 °C (programmed)
  • ≤0.32 mm I.D., ≥2 μm: -60 °C to 300 °C (isothermal or programmed)
  • ≥0.53 mm I.D., <2 μm: -60 °C to 300 °C (isothermal) or 320 °C (programmed)
  • ≥0.53 mm I.D., ≥2 μm: -60 °C to 260 °C (isothermal) or 280 °C (programmed)
기타 정보
We offer a variety of chromatography accessories including analytical syringes
법적 정보
ASTM is a registered trademark of American Society for Testing and Materials
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)