문서
SBP3500396
Sigma-Aldrich
ESX1 Blocking Peptide for SAB3500396
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12352203
NACRES:
NA.41
생물학적 소스
synthetic
Quality Level
형태
liquid
종 반응성
human
농도
200 μg/mL
배송 상태
wet ice
저장 온도
−20°C
면역원
19 amino acids near the amino terminus of human ESX1.
애플리케이션
Used as a blocking peptide in immunoblotting applications.
결합
This peptide blocks the action of antibody SAB3500396.
면책조항
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable