양식
powder
색상
white
solubility
DMSO: 4 mg/mL
H2O: 5 mg/mL
SMILES string
NC(C(O)=O)c1cccc(O)c1
생화학적/생리학적 작용
Metabotropic glutropic glutamate receptor agonist possessing some selectivity for mGluR1.
주의사항
Hygroscopic, photosensitive
신호어
Warning
유해 및 위험 성명서
예방조치 성명서
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
개인 보호 장비
dust mask type N95 (US), Eyeshields, Gloves
가장 최신 버전 중 하나를 선택하세요:
I Brabet et al.
Neuropharmacology, 34(8), 895-903 (1995-08-01)
The effects of the phenylglycine derivatives, alpha-methyl-4-carboxyphenylglycine (MCPG), 4-carboxyphenylglycine (4CPG), 4-carboxy-3-hydroxyphenylglycine (4C3HPG), 3-hydroxyphenylglycine (3HPG) and 3,5-dihydrohyphenylglycine (DHPG) were tested on LLC-PK1 cells transiently expressing the rat mGluR1a or mGluR5a receptors. As previously reported by others, (S)-3HPG and (RS)-DHPG were found
A Buisson et al.
Neuropharmacology, 34(8), 1081-1087 (1995-08-01)
We examined the effect of two novel phenylglycine derivative drugs on excitotoxicity in murine cortical cell cultures: S-4-carboxy-3-hydroxy-phenylglycine (4C3HPG), a selective agonist of mGluRs 2/3 and an antagonist at mGluRs 1/5, and S-3 hydroxy-phenylglycine (3HPG), an agonist of mGluRs 1/5.
Y Hayashi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 14(5 Pt 2), 3370-3377 (1994-05-01)
The metabotropic glutamate receptors (mGluRs) consist of at least seven different subtypes and are coupled to intracellular signal transduction via G proteins. However, the lack of specific antagonists for the mGluRs limited the precise characterization of the role of the
Wannan Tang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(7), 3016-3021 (2015-02-24)
To date, it has been difficult to reveal physiological Ca(2+) events occurring within the fine astrocytic processes of mature animals. The objective of the study was to explore whether neuronal activity evokes astrocytic Ca(2+) signals at glutamatergic synapses of adult
자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..
고객지원팀으로 연락바랍니다.