주요 문서
GE80-6444-23
Blot processing tray pack for Processor Plus
pack of 1 kit
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.22
특징
wetted part: no
포장
pack of 1 kit
제조업체/상표
Cytiva 80644423
파라미터
4-40 °C temp. range
일반 설명
Blot Processing Tray Pack
Processor Plus is a novel fluid delivery system that automates acrylamide gel staining and western blot processing. Changing from one application to the other is achieved by a fast, simple switch of a tray.
Storage Class Code
13 - Non Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)