주요 문서
BMSH3565180LCI
Sigma-Aldrich
Benchmark IncuView Integrated Microscope Shelf
for 180L CO2 Incubators (H3565-180, 180HD, 180HDO2 only)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104510
NACRES:
NC.52
특징
for 180L CO2 Incubators (H3565-180, 180HD, 180HDO2 only)
포장
pack of 1 ea
제조업체/상표
Benchmark Scientific H3565-180-LCI
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The (patent pending) IncuView LCI (Live Cell Imaging) technology for viewing cells in “real time” is an internal microscope and large, external display that allows cells to be viewed without removal from the incubator, eliminating the inherent temperature fluctuations and risk of contamination. Cells are easily viewed on the large, integrated display or viewed locally or remotely with a wifi enabled phone, tablet or computer. Cell viewing requires the IncuView Integrated Microscope Shelf.
특징 및 장점
Live cell imaging of samples in incubator, remote viewing
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)