문서
Z231037
Sigma-Aldrich
Aldrich® Library of 13C and 1H FT-NMR Spectra
C. Pouchert and J. Behnke, Aldrich Chemical Co., 1993, 4300 pp., hard cover
로그인조직 및 계약 가격 보기
About This Item
ISBN-10:
0-94163334-9
ISBN-13:
978-0-94163334-5
UNSPSC 코드:
55101509
출판물 정보
C. Pouchert and J. Behnke, Aldrich Chemical Co., 1993, 4300 pp., hard cover
일반 설명
This three volume set of 12,000 high-resolution 75MHz 13C and 300MHz 1H FT-NMR spectra is arranged according to functionality.
법적 정보
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC