형태
honeycomb
제조업체/상표
Goodfellow 995-686-25
셀 크기
8 mm
크기
300 × 300 mm
두께
10 mm
일반 설명
For updated SDS information please visit www.goodfellow.com
법적 정보
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable