Introduction to Gold Nanoparticles – Properties & Applications in Life Science
Benny Pacheco, Ph.D.
Lund University, Sweden
Overview
Gold nanoparticles are particles with diameters in the 1-100 nm range and have unique optical and physical properties, the most pronounced being the intense absorbance and scattering of incident light at its surface plasmon resonance wavelength.
The optical properties of gold nanoparticles are governed by their morphology, i.e. size, shape and aggregation status. By precisely engineering particles with different morphologies and surface chemistries, these particles can be tuned to suit a variety of applications making them a very versatile research and diagnostic tool. For example, the gold nanoparticle surface can readily be functionalized with peptides, proteins and antibodies providing them with specificity toward cellular targets both in vitro and in vivo. In addition, modifying the surface with oligonucleotides enables them to be used for genetic detection. Common applications include their use in immunohistochemistry, bioimaging, biosensors, lateral flow assays, as cellular probes, and as vehicles for drug delivery.
Areas Covered in the Webinar
- Physical and optical properties of gold nanoparticles
- Surface modification strategies and the generation of biological probes
- Detection of gold nanoparticles and their biomedical applications
Who Should Watch?
- Materials Scientists
- Biomedical Engineers
- Chemical Engineers
- Translational researcher