Simplified Olefin Isomerization by Cobalt/Silane
Introduction
Recently, the Shenvi lab has repurposed Jacobsen’s cobalt chloride complex, Co(salen,t-Bu, t-Bu)Cl, to enable positional isomerization of terminal alkenes using reversible hydrogen atom transfer (HAT). This cobalt pre-catalyst, when combined with a catalytic amount of organosilane, enables high-yielding isomerizations and cycloisomerizations of alkenes in the presence of strongly Lewis basic functional groups like tertiary amines and imidazoles. The Shenvi group of The Scripps Research Institute and Chemistry now offer the inexpensive, bench-top stable Co(salen,t-Bu, t-Bu)Cl complex (804010) that can be readily used outside the glovebox.
Co(salen,t-Bu, t-Bu)Cl complex (804010)
Application Scope
Advantages
- Functional group tolerance
- Diastereoselective isomerization
- Prevents reduction
- No need for a glove box
- High-yielding, high-profiting and low-cost reactions (Scheme 2)
Representative Schemes
Materials
Loading
References
1.
Crossley SWM, Barabé F, Shenvi RA. 2014. Simple, Chemoselective, Catalytic Olefin Isomerization. J. Am. Chem. Soc.. 136(48):16788-16791. https://doi.org/10.1021/ja5105602
2.
Iwasaki K, Wan KK, Oppedisano A, Crossley SWM, Shenvi RA. 2014. Simple, Chemoselective Hydrogenation with Thermodynamic Stereocontrol. J. Am. Chem. Soc.. 136(4):1300-1303. https://doi.org/10.1021/ja412342g
3.
King SM, Ma X, Herzon SB. 2014. A Method for the Selective Hydrogenation of Alkenyl Halides to Alkyl Halides. J. Am. Chem. Soc.. 136(19):6884-6887. https://doi.org/10.1021/ja502885c
4.
Shigehisa H, Aoki T, Yamaguchi S, Shimizu N, Hiroya K. 2013. Hydroalkoxylation of Unactivated Olefins with Carbon Radicals and Carbocation Species as Key Intermediates. J. Am. Chem. Soc.. 135(28):10306-10309. https://doi.org/10.1021/ja405219f
5.
Mukaiyama T, Yamada T. 1995. Recent Advances in Aerobic Oxygenation. BCSJ. 68(1):17-35. https://doi.org/10.1246/bcsj.68.17
6.
Gaspar B, Carreira E. 2008. Catalytic Hydrochlorination of Unactivated Olefins withpara?Toluenesulfonyl Chloride. Angew. Chem. Int. Ed.. 47(31):5758-5760. https://doi.org/10.1002/anie.200801760