Merck
HomeOrganic Reaction ToolboxPeriodic Table of the Elements
Periodic Table of the Elements
The periodic table (also known as the periodic table of elements) is organized so scientists can quickly discern the properties of individual elements such as their mass, electron number, electron configuration and their unique chemical properties. Metals reside on the left side of the table, while non-metals reside on the right. Organizing the elements to help further our understanding was first provided by Dmitri Mendeleev.
This periodic table of the elements with names, atomic number, symbol and mass is color-coded for easier reference by students and researchers. For quick reference, go to the periodic table chart with names listed alphabetical order. 
Periodic table of the elements
Click the image to download and view the periodic table of the elements.
Periodic Table with Element Names and Electronegativity
This periodic table chart lists elements by name in alphabetical order including the element symbol, atomic number, and Pauling electronegativity value for quick and simple reference.
Element NameSymbolAtomic NumberElectronegativity (χ)
ActiniumAc891.1
AluminumAl131.61
AmericiumAm951.3
AntimonySb512.05
ArgonAr18
ArsenicAs332.18
AstatineAt852.2
BariumBa560.89
BerkeliumBk971.3
BerylliumBe41.57
BismuthBi832.02
BohriumBh107
BoronB52.04
BromineBr352.96
CadmiumCd481.69
CalciumCa201
CaliforniumCf981.3
CarbonC62.55
CeriumCe581.12
CesiumCs550.79
ChlorineCl173.16
ChromiumCr241.66
CobaltCo271.88
CopperCu291.9
CuriumCm961.3
DarmstadtiumDs110
DubniumDb105
DysprosiumDy661.22
EinsteiniumEs991.3
ErbiumEr681.24
EuropiumEu63
FermiumFm1001.3
FluorineF93.98
FranciumFr870.7
GadoliniumGd641.2
GalliumGa311.81
GermaniumGe322.01
GoldAu792.54
HafniumHf721.3
HassiumHs108
HeliumHe2
HolmiumHo671.23
HydrogenH12.2
IndiumIn491.78
IodineI532.66
IridiumIr772.2
IronFe261.83
KryptonKr363
LanthanumLa571.1
LawrenciumLr103
LeadPb822.33
LithiumLi30.98
LutetiumLu711.27
MagnesiumMg121.31
ManganeseMn251.55
MeitneriumMt109
MendeleviumMd1011.3
MercuryHg802
MolybdenumMo422.16
NeodymiumNd601.14
NeonNe10
NeptuniumNp931.36
NickelNi281.91
NiobiumNb411.6
NitrogenN73.04
NobeliumNo1021.3
OganessonUuo118
OsmiumOs762.2
OxygenO83.44
PalladiumPd462.2
PhosphorusP152.19
PlatinumPt782.28
PlutoniumPu941.28
PoloniumPo842
PotassiumK190.82
PraseodymiumPr591.13
PromethiumPm61
ProtactiniumPa911.5
RadiumRa880.9
RadonRn86
RheniumRe751.9
RhodiumRh452.28
RoentgeniumRg111
RubidiumRb370.82
RutheniumRu442.2
RutherfordiumRf104
SamariumSm621.17
ScandiumSc211.36
SeaborgiumSg106
SeleniumSe342.55
SiliconSi141.9
SilverAg471.93
SodiumNa110.93
StrontiumSr380.95
SulfurS162.58
TantalumTa731.5
TechnetiumTc431.9
TelluriumTe522.1
TerbiumTb65
ThalliumTl811.62
ThoriumTh901.3
ThuliumTm691.25
TinSn501.96
TitaniumTi221.54
TungstenW742.36
UnunbiumUub112 
UnunhexiumUuh116 
UnunpentiumUup115 
UnunquadiumUuq114 
UnunseptiumUus117 
UnuntriumUut113 
UraniumU921.38
VanadiumV231.63
XenonXe542.6
YtterbiumYb70
YttriumY391.22
ZincZn301.65
ZirconiumZr401.33