HPLC Analysis of Fumaric, Oxalic, and Citric Acid on SeQuant™ ZIC-HILIC
Materials
accessory
Product No.
Description
Pricing
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),coated tip
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),coated tip
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate
1.5 mL/min
pressure
1240 psi (8.6 MPa)
detector
220 nm
injection
20 μL
Description
Legal Information
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany