Merck
HomePhotometry & ReflectometryAnalytical Method: Analytical Quality Assurance, Standard for Free Chlorine (DIN)
Analytical Method: Analytical Quality Assurance, Standard for Free Chlorine (DIN)
Reagents:
Potassium iodate volumetric standard (Product No. 102404)
Potassium iodide (Product No. 105043) for analysis
Sulfuric acid (Product No. 109072 ) 0.5 mol/L
Sodium hydroxide solution (Product No. 109136) 2 mol/L
Water (Product No. 116754) for analysis
Preparation of a KlO3 stock solution:
Dissolve 1.006 g of KlO3 in 250 mL of water for analysis in a calibrated or conformity-checked 1,000-mL volumetric flask. Subsequently make up to the mark with water for analysis.
Preparation of a KlO3/Kl standard solution:
Transfer 15.0 mL of the KlO3 stock solution to a calibrated or conformity-checked 1,000-mL volumetric flask, add approx. 1 g of Kl and make up to the mark with water for analysis. 1 mL of this solution is equivalent to 0.015 mg of free chlorine.
Preparation of the chlorine standard solution:
Pipette 20.0 mL KlO3/Kl standard solution into a calibrated or conformity-checked 100-mL volumetric flask, add
2 mL of H2SO4 0.5 mol/L, leave to stand for 1 min, and then add NaOH 2 mol/L dropwise (approx. 1 ml) until the solution just loses its color. Subsequently make up the solution to the mark with water for analysis. The concentration of the solution is 3.00 mg/L free chlorine.
Stability:
The KlO3 stock solution remains stable for 4 weeks when stored in a cool place. The KlO3/Kl standard solution can be used for 5 hours when stored in a cool place. The dilute iodine standard solution is not stable and must be used immediately.
For use with test kits:
Measuring range
in mg/L Cl2
117925
MQuantTM Chlorine Test (free chlorine)
4 - 120
117924
MQuantTM Chlorine Test (free chlorine)
25 - 500
114434
MColortestTM Chlorine Test (free chlorine)
0.01 - 0.3
111174
MColortestTM Chlorine and pH Test (free + total chlorine)
0.1 - 1.5
111157
MColortestTM Chlorine and pH Test (free + total chlorine) refill pack for 111174
111113
MColortestTM Chlorine Test Reagent Cl2-1
111114
MColortestTM Chlorine Test Reagent Cl2-2
111160
MColortestTM Chlorine and pH Test (free chlorine)
0.1 - 1.5
114670
MColortestTM Chlorine Test (free chlorine)
0.1 - 2
114978
MColortestTM Chlorine Test (free chlorine, liquid)
0.1 - 2
114979
MColortestTM Chlorine Test (free chlorine, liquid)
refill pack for 114978
1.14801
MColortestTM Chlorine Test (free + total chlorine, liquid)
0.1 - 2
114803
MColortestTM Chlorine Test (free + total chlorine, liquid)
refill pack for 114801
114976
MColortestTM Chlorine Test (free chlorine)
0.25 - 15
114977
MColortestTM Chlorine Test (free chlorine)
refill pack for 114976
114826
MColortestTM Chlorine Test (free + total chlorine)
0.25 - 15
118326
MColortestTM Chlorine Test (free + total chlorine)
refill pack for 114826
116896
Reflectoquant® Chlorine Test (free + total chlorine)
0.5 - 10.0
100595
Spectroquant® Chlorine Cell Test (free chlorine)
0.03 - 6.00
100598
Spectroquant® Chlorine Test (free chlorine)
0.010 - 6.00
100086
Spectroquant® Chlorine Reagents Cl2-1
0.010 - 6.00
100087
Spectroquant® Chlorine Reagents Cl2-2
0.010 - 6.00
119254
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 10-ml-samples (free chlorine)
0 - 2.00
119256
Spectroquant® Chlorine Powder Packs for non-Merck photometers for 25-ml-samples (free chlorine)
0 - 10.00
100597
Spectroquant® Chlorine Cell Test (free + total chlorine)
0.03 - 6.00
100599
Spectroquant® Chlorine Test (free + total chlorine)
0.010 - 6.00
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch