Merck
HPLC Analysis of the Peptide Glycine-Leucine-Alanine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of the Peptide Glycine-Leucine-Alanine Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] 5 mM ammonium acetate, pH 4.1; [B] acetonitrile; (35:65, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC