Merck
HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
HPLC Analysis of Phenylalanine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
guard cartridge
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Guard Column
Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
standard
Product No.
Description
Pricing
L-Phenylalanine
from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
L-Phenylalanine
reagent grade, ≥98%
D-Phenylalanine
≥98% (HPLC)
L-Phenylalanine
BioUltra, ≥99.0% (NT)
L-Phenylalanine
99%, FCC
L-Phenylalanine
analytical standard, for Nitrogen Determination According to Kjeldahl Method
L-Phenylalanine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] methanol; [C] formic acid; (30:70:0.02, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
300 μg /mL in 30:70 water:methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC