Merck
GC Analysis of FAMEs in Human Plasma on Equity-1
GC Analysis of FAMEs in Human Plasma on Equity-1 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Equity-1, 30 m x 0.25 mm I.D., x 0.25 μm (28046-U)
column
SUPELCOWAX 10, 0.95 m x 0.10 mm x 0.10 μm
oven
180 °C, 3 °C/min to 280 °C (10 min)
detector
FID
carrier gas
hydrogen, 75.3 kPa, constant
Description
Legal Information
null