- Suppression of homology-dependent DNA double-strand break repair induces PARP inhibitor sensitivity in
Suppression of homology-dependent DNA double-strand break repair induces PARP inhibitor sensitivity in
Oncotarget (2018-02-13)
Susan E Scanlon, Denise C Hegan, Parker L Sulkowski, Peter M Glazer
PMID29435132
ABSTRACT
The von Hippel-Lindau (